Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163792.1 | NP_001157264.1 | 1, 2 | 202 | |
NM_145553.2 | NP_663528.1 | 1, 2 | 202 | ||
GenBank mRNA | AK038831.1 | - | 1, 2 | 210 | |
AK154575.1 | - | 1, 2 | 203 | ||
BC008163.1 | - | 1, 2 | 162 | 3709256 |