Gene Transcripts
- Gene Symbol
- Atg13
- Entrez Gene ID
- 51897
- Gene Name
- autophagy related 13
- Gene Aliases
- 1110053A20Rik, D2Ertd391e, Harbi1
- Chromosome Location
- Chr. 2: 91674612 - 91710598 on Build GRCm38
- UniGene ID
- Mm.28492
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145528.3 |
NP_663503.1
|
4, 5 |
637 |
|
GenBank mRNA |
AK149672.1 |
- |
4, 5 |
596 |
|
AK154470.1 |
- |
4, 5 |
623 |
|
AK163962.1 |
- |
4, 5 |
589 |
|
AK171206.1 |
- |
4, 5 |
623 |
|
AK171440.1 |
- |
4, 5 |
614 |
|
BC016669.1 |
- |
4, 5 |
621 |
2648797 |
BC023673.1 |
- |
4, 5 |
587 |
5343236 |
BC023712.1 |
- |
4, 5 |
610 |
5346303 |
BC033419.1 |
- |
4, 5 |
588 |
5359944 |
View Full Product Details