Gene Transcripts

Gene Symbol
Slc5a9
Entrez Gene ID
230612
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 9
Gene Aliases
AI159731, SGLT4
Chromosome Location
Chr. 4: 111875374 - 111902796 on Build GRCm38
UniGene ID
Mm.26630
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145551.4 NP_663526.3 8 981
GenBank mRNA AK029158.1 - 8 982
AK033425.1 - 7 911
AK050696.1 - 7 861
BC021357.1 - 8 962 5012356

View Full Product Details