Gene Transcripts

Gene Symbol
Krcc1
Entrez Gene ID
57896
Gene Name
lysine-rich coiled-coil 1
Gene Aliases
AA792894
Chromosome Location
Chr. 6: 71271569 - 71285319 on Build GRCm38
UniGene ID
Mm.250569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145568.3 NP_663543.1 4 587
GenBank mRNA AK050640.1 - 3 515
AK146818.1 - 3 526
AK148096.1 - 3 505
AK162678.1 - 3 515
BC005704.1 - 2 232 3598565
BC046798.1 - 3 483 6403577
BC069863.1 - 4 587 6306896

View Full Product Details