Gene Transcripts

Gene Symbol
Vmn1r69
Entrez Gene ID
252904
Gene Name
vomeronasal 1 receptor 69
Gene Aliases
V1re9
Chromosome Location
Chr. 7: 10579756 - 10581487 on Build GRCm38
UniGene ID
Mm.222635
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145842.3 NP_665841.1 1 1070
GenBank mRNA AF394954.1 - 1 1069

View Full Product Details