Gene Transcripts

Gene Symbol
Lctl
Entrez Gene ID
235435
Gene Name
lactase-like
Gene Aliases
E130104I05Rik, KLPH
Chromosome Location
Chr. 9: 64117123 - 64138118 on Build GRCm38
UniGene ID
Mm.436581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145835.2 NP_665834.1 12 1674
GenBank mRNA AF309072.1 - 12 1675
AK053515.1 - 11 1509
BC030631.1 - 8 999 1195880

View Full Product Details