Gene Transcripts

Gene Symbol
Tspear
Entrez Gene ID
252974
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C330046G03Rik, ORF65, TSP-EAR, Tnep1
Chromosome Location
Chr. 10: 77686030 - 77886994 on Build GRCm38
UniGene ID
Mm.221237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287074.1 NP_001274003.1 5 1026
GenBank mRNA AJ487520.1 - 4 697
AK082868.1 - 3 498
JQ815565.1 - 5 1026

View Full Product Details