Gene Transcripts

Gene Symbol
Slc26a8
Entrez Gene ID
224661
Gene Name
solute carrier family 26, member 8
Gene Aliases
Chromosome Location
Chr. 17: 28637779 - 28691237 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290320.1 NP_001277249.1 2 383
GenBank mRNA BC027076.1 - 2 383

View Full Product Details