Gene Transcripts
- Gene Symbol
- NME7
- Entrez Gene ID
- 29922
- Gene Name
- NME/NM23 family member 7
- Gene Aliases
- CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
- Chromosome Location
- Chr. 1: 169132385 - 169367963 on Build GRCh38
- UniGene ID
- Hs.706952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013330.4 |
NP_037462.1
|
4, 5 |
650 |
|
NM_197972.2 |
NP_932076.1
|
4, 5 |
619 |
|
NR_104229.1 |
|
4, 5 |
650 |
|
XM_005245106.4 |
XP_005245163.1
|
2, 3 |
228 |
|
XM_017001099.1 |
XP_016856588.1
|
4, 5 |
636 |
|
XM_017001100.1 |
XP_016856589.1
|
2, 3 |
216 |
|
GenBank mRNA |
AB209049.1 |
- |
4, 5 |
459 |
|
AF153191.2 |
- |
4, 5 |
489 |
|
AK094513.1 |
- |
4, 5 |
602 |
|
AK290701.1 |
- |
4, 5 |
484 |
|
AK301917.1 |
- |
4, 5 |
633 |
|
BC006983.1 |
- |
4, 5 |
481 |
3997152 |
CD107958.1 |
- |
4, 5 |
678 |
|
View Full Product Details