Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146218.4 | NP_666330.2 | 2 | 231 | |
GenBank mRNA | AK137499.1 | - | 2 | 211 | |
AK137698.1 | - | 2 | 194 | ||
AK139032.1 | - | 2 | 211 | ||
AK146451.1 | - | 2 | 201 | ||
BC023694.1 | - | 2 | 236 | 5344528 | |
BC096602.1 | - | 2 | 194 | 4954068 |