Gene Transcripts

Gene Symbol
Tbc1d13
Entrez Gene ID
70296
Gene Name
TBC1 domain family, member 13
Gene Aliases
2600014A06Rik, BC025586
Chromosome Location
Chr. 2: 30133751 - 30152013 on Build GRCm38
UniGene ID
Mm.19172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146252.2 NP_666364.1 5 302
GenBank mRNA AK158066.1 - 5 383
BC025586.1 - 5 328 5324875

View Full Product Details