Gene Transcripts
- Gene Symbol
- RPL34
- Entrez Gene ID
- 6164
- Gene Name
- ribosomal protein L34
- Gene Aliases
- L34
- Chromosome Location
- Chr. 4: 108620558 - 108630484 on Build GRCh38
- UniGene ID
- Hs.438227
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000995.4 |
NP_000986.2
|
4, 5 |
329 |
|
NM_001319232.1 |
NP_001306161.1
|
5, 6 |
475 |
|
NM_001319234.1 |
NP_001306163.1
|
4, 5 |
459 |
|
NM_001319235.1 |
NP_001306164.1
|
4, 5 |
409 |
|
NM_001319236.1 |
NP_001306165.1
|
4, 5 |
329 |
|
NM_033625.3 |
NP_296374.1
|
4, 5 |
409 |
|
GenBank mRNA |
AA128058.1 |
- |
5, 6 |
434 |
|
AF147334.1 |
- |
3, 4 |
233 |
|
AY762981.1 |
- |
1, 2 |
112 |
|
BC001773.1 |
- |
4, 5 |
343 |
3355603 |
BC070208.1 |
- |
4, 5 |
310 |
6724916 |
BC106009.1 |
- |
4, 5 |
321 |
4390762 |
BE394007.1 |
- |
4, 5 |
375 |
|
BU588998.1 |
- |
4, 5 |
409 |
|
CB132938.1 |
- |
4, 5 |
460 |
|
CD175155.1 |
- |
4, 5 |
431 |
|
CD249923.1 |
- |
4, 5 |
367 |
|
CR542232.1 |
- |
3, 4 |
277 |
|
CR542242.1 |
- |
3, 4 |
277 |
|
DA779124.1 |
- |
4, 5 |
310 |
|
L38941.1 |
- |
3, 4 |
297 |
|
View Full Product Details