Gene Transcripts

Gene Symbol
Ly6g5b
Entrez Gene ID
266614
Gene Name
lymphocyte antigen 6 complex, locus G5B
Gene Aliases
Chromosome Location
Chr. 17: 35113944 - 35115400 on Build GRCm38
UniGene ID
Mm.104680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148939.2 NP_683741.1 3 259
GenBank mRNA AJ315553.1 - 3 259
AJ315554.1 - 2 355
AK151902.1 - 2 296
BC131964.1 - 3 287 40130241
BC131966.1 - 3 287 40130243
BC145150.1 - 2 383
HE864438.1 - 3 275
HE864439.1 - 2 371

View Full Product Details