Gene Transcripts
- Gene Symbol
- Slc44a2
- Entrez Gene ID
- 68682
- Gene Name
- solute carrier family 44, member 2
- Gene Aliases
- 1110028E10Rik, CTL2
- Chromosome Location
- Chr. 9: 21320719 - 21355028 on Build GRCm38
- UniGene ID
- Mm.148425
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199186.1 |
NP_001186115.1
|
2, 3 |
174 |
|
NM_152808.3 |
NP_690021.1
|
2, 3 |
176 |
|
GenBank mRNA |
AK029457.1 |
- |
2, 3 |
136 |
|
AK041533.1 |
- |
2, 3 |
133 |
|
AK043924.1 |
- |
2, 3 |
146 |
|
AK048648.1 |
- |
2, 3 |
161 |
|
AK088553.1 |
- |
2, 3 |
123 |
|
AK153992.1 |
- |
2, 3 |
155 |
|
AK166695.1 |
- |
2, 3 |
177 |
|
BC023763.1 |
- |
2, 3 |
159 |
|
BC031535.1 |
- |
2, 3 |
152 |
3991609 |
BY339157.1 |
- |
2, 3 |
176 |
|
View Full Product Details