Gene Transcripts

Gene Symbol
Las1l
Entrez Gene ID
76130
Gene Name
LAS1-like (S. cerevisiae)
Gene Aliases
1810030A06Rik, 5830482G23Rik
Chromosome Location
Chr. X: 95920045 - 95956974 on Build GRCm38
UniGene ID
Mm.274318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152822.3 NP_690035.2 2, 3 451
GenBank mRNA AK018059.1 - 2, 3 381
AK089160.2 - 2, 3 403
AK163919.1 - 2, 3 440
BC004735.1 - 1, 2 43
BC032218.1 - 2, 3 374
BC141155.1 - 2, 3 388 9055767
BQ573371.1 - 1, 2 31

View Full Product Details