Gene Transcripts

Gene Symbol
Osbpl10
Entrez Gene ID
74486
Gene Name
oxysterol binding protein-like 10
Gene Aliases
4933433D06Rik, C820004B04Rik, OPR-10
Chromosome Location
Chr. 9: 114978583 - 115233273 on Build GRCm38
UniGene ID
Mm.458474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148958.2 NP_683761.1 6 874
GenBank mRNA AK017034.1 - 8 1205
AK037617.1 - 5 1015
BC027256.1 - 6 874 2650570

View Full Product Details