Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153088.2 | NP_694728.1 | 7 | 1886 | |
GenBank mRNA | AK089478.1 | - | 7 | 1794 | |
AK167390.1 | - | 7 | 1644 | ||
AY028804.1 | - | 7 | 1911 | ||
BC049184.1 | - | 7 | 1746 | 6439402 | |
BC058085.1 | - | 6 | 2073 | ||
BC065158.1 | - | 7 | 1621 | 5717102 | |
BC079638.1 | - | 7 | 1621 | 5717102 |