Gene Transcripts
- Gene Symbol
- Ttc39a
- Entrez Gene ID
- 230603
- Gene Name
- tetratricopeptide repeat domain 39A
- Gene Aliases
- 4922503N01Rik
- Chromosome Location
- Chr. 4: 109394148 - 109444745 on Build GRCm38
- UniGene ID
- Mm.33091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145948.1 |
NP_001139420.1
|
3, 4 |
346 |
|
NM_153392.2 |
NP_700441.2
|
3, 4 |
355 |
|
GenBank mRNA |
AK048659.1 |
- |
3, 4 |
356 |
|
AK076641.1 |
- |
3, 4 |
308 |
|
AK076720.1 |
- |
3, 4 |
347 |
|
AK076772.1 |
- |
3, 4 |
309 |
|
AK082733.1 |
- |
3, 4 |
346 |
|
AK172951.1 |
- |
3, 4 |
278 |
|
BC023883.1 |
- |
3, 4 |
354 |
5150841 |
BC026660.1 |
- |
3, 4 |
296 |
4988900 |
BY095692.1 |
- |
3, 4 |
363 |
|
View Full Product Details