Gene Transcripts

Gene Symbol
DAPK2
Entrez Gene ID
23604
Gene Name
death associated protein kinase 2
Gene Aliases
DRP-1, DRP1
Chromosome Location
Chr. 15: 63907036 - 64046470 on Build GRCh38
UniGene ID
Hs.237886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014326.3 NP_055141.2 12 2401
GenBank mRNA AK026801.1 - 1 1943

View Full Product Details