Gene Transcripts

Gene Symbol
Pygb
Entrez Gene ID
110078
Gene Name
brain glycogen phosphorylase
Gene Aliases
Chromosome Location
Chr. 2: 150786796 - 150831748 on Build GRCm38
UniGene ID
Mm.222584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153781.1 NP_722476.1 3 464
GenBank mRNA AK032769.1 - 3 477
AK035673.1 - 3 193
AK134667.1 - 3 474
AK142716.1 - 3 473
AK147763.1 - 3 496
AK168381.1 - 3 474
AK169052.1 - 3 499
AK169631.1 - 3 478
BC031457.1 - 3 472
BC035283.1 - 3 464 4947455

View Full Product Details