Gene Transcripts
- Gene Symbol
- Tmem18
- Entrez Gene ID
- 211986
- Gene Name
- transmembrane protein 18
- Gene Aliases
- BC024093
- Chromosome Location
- Chr. 12: 30584443 - 30591219 on Build GRCm38
- UniGene ID
- Mm.34035
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC024093.1 |
- |
5 |
1912 |
|
View Full Product Details