Gene Transcripts

Gene Symbol
Dio3
Entrez Gene ID
107585
Gene Name
deiodinase, iodothyronine type III
Gene Aliases
Chromosome Location
Chr. 12: 110279230 - 110281097 on Build GRCm38
UniGene ID
Mm.154427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172119.2 NP_742117.2 1 1709
GenBank mRNA AK140797.1 - 1 1710
AK146060.1 - 1 1710

View Full Product Details