Gene Transcripts

Gene Symbol
Mrpl21
Entrez Gene ID
353242
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
BC028768
Chromosome Location
Chr. 19: 3282920 - 3292837 on Build GRCm38
UniGene ID
Mm.74084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172252.3 NP_758456.2 6 575
GenBank mRNA AK003285.1 - 6 562
AK020739.1 - 6 576
BC028768.1 - 6 550 1282313

View Full Product Details