Gene Transcripts
- Gene Symbol
- Exog
- Entrez Gene ID
- 208194
- Gene Name
- endo/exonuclease (5'-3'), endonuclease G-like
- Gene Aliases
- AW557704, ENGL-B, ENGL-a, Endogl1, Endogl2, Engl, Engla, Englb
- Chromosome Location
- Chr. 9: 119444923 - 119465518 on Build GRCm38
- UniGene ID
- Mm.101578
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172136.1 |
NP_001165607.1
|
3, 4 |
408 |
|
NM_172456.3 |
NP_766044.1
|
2, 3 |
373 |
|
GenBank mRNA |
AK028887.1 |
- |
2, 3 |
337 |
|
AK034654.1 |
- |
3, 4 |
370 |
|
AK051300.1 |
- |
3, 4 |
372 |
|
AK148426.1 |
- |
2, 3 |
354 |
|
AK162566.1 |
- |
2, 3 |
336 |
|
AK163555.1 |
- |
2, 3 |
337 |
|
BC059920.1 |
- |
3, 4 |
351 |
|
BC064730.1 |
- |
3, 4 |
423 |
|
BC119024.1 |
- |
2, 3 |
337 |
8733773 |
BC144729.1 |
- |
3, 4 |
372 |
|
View Full Product Details