Gene Transcripts

Gene Symbol
Bzrap1
Entrez Gene ID
207777
Gene Name
benzodiazepine receptor associated protein 1
Gene Aliases
D230016K05, PRAX-1, mKIAA0612
Chromosome Location
Chr. 11: 87759066 - 87785928 on Build GRCm38
UniGene ID
Mm.342019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172449.2 NP_766037.2 27 6078
GenBank mRNA AK051895.1 - 6 2027
AK122329.1 - 5 949
BC055691.1 - 7 829

View Full Product Details