Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167976.1 | NP_001161448.1 | 11 | 1198 | |
NM_172577.3 | NP_766165.2 | 10 | 1641 | ||
GenBank mRNA | AK036988.1 | - | 10 | 1380 | |
AK041694.1 | - | 10 | 1642 | ||
AK045180.1 | - | 10 | 1466 | ||
AK162379.1 | - | 10 | 1467 | ||
AK167535.1 | - | 10 | 1349 | ||
BC057980.1 | - | 10 | 1465 | 5042653 | |
EU180236.1 | - | 11 | 1198 |