Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134335.1 | NP_001127807.1 | 4 | 577 | |
NM_014413.3 | NP_055228.2 | 4 | 577 | ||
GenBank mRNA | AB037790.1 | - | 4 | 502 | |
AF100784.1 | - | 4 | 466 | ||
AF147094.1 | - | 4 | 512 | ||
AF183414.1 | - | 4 | 464 | ||
AF255050.1 | - | 4 | 522 | ||
AK075192.1 | - | 4 | 505 | ||
AK290327.1 | - | 4 | 510 | ||
AK295712.1 | - | 3 | 378 | ||
AL136563.1 | - | 4 | 482 | ||
AL834494.1 | - | 4 | 482 | ||
BC006524.2 | - | 4 | 509 | 2989989 | |
DC341983.1 | - | 4 | 577 |