Gene Transcripts
- Gene Symbol
- Rab2b
- Entrez Gene ID
- 76338
- Gene Name
- RAB2B, member RAS oncogene family
- Gene Aliases
- 1500012D09Rik, 4930528G15Rik, A230002G14, D530043M21Rik
- Chromosome Location
- Chr. 14: 52261759 - 52279395 on Build GRCm38
- UniGene ID
- Mm.32870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172601.3 |
NP_766189.1
|
6, 7 |
636 |
|
GenBank mRNA |
AB232586.1 |
- |
6, 7 |
482 |
|
AK005230.1 |
- |
6, 7 |
539 |
|
AK038393.1 |
- |
6, 7 |
571 |
|
AK044202.1 |
- |
6, 7 |
513 |
|
AK160249.1 |
- |
6, 7 |
787 |
|
AK160351.1 |
- |
6, 7 |
502 |
|
BC033312.1 |
- |
6, 7 |
533 |
4481516 |
BC046334.1 |
- |
6, 7 |
578 |
6492838 |
BC054719.1 |
- |
6, 7 |
569 |
|
BC061513.1 |
- |
2, 3 |
585 |
|
View Full Product Details