Gene Transcripts

Gene Symbol
Mthfsd
Entrez Gene ID
234814
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
AW049977, BC052066, C920004D05
Chromosome Location
Chr. 8: 121097555 - 121108409 on Build GRCm38
UniGene ID
Mm.31690
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166482.1 NP_001159954.1 5, 6 523
NM_172761.3 NP_766349.1 5, 6 617
GenBank mRNA AK083317.1 - 5, 6 557
AK141278.1 - 5, 6 452
BC023782.1 - 5, 6 594
BC026652.1 - 4, 5 493

View Full Product Details