Gene Transcripts

Gene Symbol
Shisa7
Entrez Gene ID
232813
Gene Name
shisa family member 7
Gene Aliases
Chromosome Location
Chr. 7: 4825551 - 4844789 on Build GRCm38
UniGene ID
Mm.44666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290291.1 NP_001277220.1 5 1950
NM_172737.4 NP_766325.3 6 2001
GenBank mRNA AK085127.1 - 5 1223
AK145016.1 - 4 1235

View Full Product Details