Gene Transcripts

Gene Symbol
Gucy1b2
Entrez Gene ID
239134
Gene Name
guanylate cyclase 1, soluble, beta 2
Gene Aliases
B930025B03, GC-SB2
Chromosome Location
Chr. 14: 62373023 - 62457562 on Build GRCm38
UniGene ID
Mm.236557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204340.1 NP_001191269.1 2 241
NM_172810.3 NP_766398.1 2 241
GenBank mRNA AK047129.1 - 2 242
BC137868.1 - 2 238 8860887
BC144988.1 - 2 238 9053536

View Full Product Details