Gene Transcripts

Gene Symbol
Pigz
Entrez Gene ID
239827
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class Z
Gene Aliases
F630022B06Rik
Chromosome Location
Chr. 16: 31926073 - 31946048 on Build GRCm38
UniGene ID
Mm.440388
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172822.3 NP_766410.2 3 1881
GenBank mRNA AK087516.1 - 3 1767
AK089183.1 - 3 1780
BC117748.2 - 2 1701 40090477

View Full Product Details