Gene Transcripts
- Gene Symbol
- Zxdc
- Entrez Gene ID
- 80292
- Gene Name
- ZXD family zinc finger C
- Gene Aliases
- A930012H20, B930086F11Rik, BC003332
- Chromosome Location
- Chr. 6: 90369483 - 90403490 on Build GRCm38
- UniGene ID
- Mm.100116
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030260.3 |
NP_084536.2
|
2, 3 |
1236 |
|
NM_173002.3 |
NP_766590.1
|
2, 3 |
1236 |
|
GenBank mRNA |
AK030287.1 |
- |
2, 3 |
1152 |
|
AK042190.1 |
- |
2, 3 |
1163 |
|
AK042392.1 |
- |
2, 3 |
1146 |
|
AK044429.1 |
- |
2, 3 |
1129 |
|
AK044529.1 |
- |
2, 3 |
1160 |
|
AK044948.1 |
- |
2, 3 |
1130 |
|
AK047079.1 |
- |
2, 3 |
1118 |
|
AK047538.1 |
- |
2, 3 |
1152 |
|
AK080307.1 |
- |
2, 3 |
1130 |
|
AK081005.1 |
- |
2, 3 |
1238 |
|
AK169517.1 |
- |
2, 3 |
1123 |
|
AK185703.1 |
- |
1, 2 |
149 |
|
AK201488.1 |
- |
1, 2 |
149 |
|
AK218430.1 |
- |
1, 2 |
149 |
|
BC003332.1 |
- |
2, 3 |
1220 |
3256958 |
View Full Product Details