Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr. 4: 109061145 - 109202272 on Build GRCm38
UniGene ID
Mm.366315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134791.2 NP_001128263.1 12 1065
NM_133885.4 NP_598646.4 15 1298
NM_173350.3 NP_775485.2 13 1104
GenBank mRNA AK035658.1 - 13 1053
AK075981.1 - 2 83
AK131984.1 - 15 1269
AK172010.1 - 13 1091
BC021507.1 - 11 926 5347121
BC023759.1 - 15 1253
BC026927.1 - 13 987 5134780
BC089163.1 - 12 1006 5367603

View Full Product Details