Gene Transcripts

Gene Symbol
Cds1
Entrez Gene ID
74596
Gene Name
CDP-diacylglycerol synthase 1
Gene Aliases
4833409J18Rik, AI314024, AW125888
Chromosome Location
Chr. 5: 101765130 - 101823858 on Build GRCm38
UniGene ID
Mm.46764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173370.3 NP_775546.2 5 802
GenBank mRNA AF533367.2 - 5 711
BC055292.1 - 5 802 6491596

View Full Product Details