Gene Transcripts

Gene Symbol
SLK
Entrez Gene ID
9748
Gene Name
STE20 like kinase
Gene Aliases
LOSK, STK2, bA16H23.1, se20-9
Chromosome Location
Chr. 10: 103967173 - 104029233 on Build GRCh38
UniGene ID
Hs.591922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304743.1 NP_001291672.1 3 906
NM_014720.3 NP_055535.2 3 906
XM_011540401.2 XP_011538703.1 3 918
GenBank mRNA AB002804.1 - 3 577
AF273048.1 - 3 379
AK302855.1 - 3 339
AK307389.1 - 4 458
BC047762.1 - 3 899
BC047885.1 - 3 906
BC064804.1 - 3 902
BC111565.1 - 3 376 40009070
D86959.1 - 3 856

View Full Product Details