Gene Transcripts

Gene Symbol
Ythdf1
Entrez Gene ID
228994
Gene Name
YTH domain family 1
Gene Aliases
2210410K23Rik, 8030473O16
Chromosome Location
Chr. 2: 180904377 - 180920936 on Build GRCm38
UniGene ID
Mm.30205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173761.3 NP_776122.1 3 312
GenBank mRNA AK033241.1 - 4 402
AK046768.1 - 3 326
AK075846.1 - 3 291
AK172582.1 - 3 286
BC061479.1 - 3 283 6334535
BC065050.1 - 3 312 6418891

View Full Product Details