Gene Transcripts

Gene Symbol
Ifnlr1
Entrez Gene ID
242700
Gene Name
interferon lambda receptor 1
Gene Aliases
CRF2-12, Il28ra
Chromosome Location
Chr. 4: 135686457 - 135708181 on Build GRCm38
UniGene ID
Mm.259623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174851.3 NP_777276.3 3 382
GenBank mRNA AK141338.1 - 3 383
AY184376.1 - 3 297
BC057856.1 - 3 315 5036481

View Full Product Details