Gene Transcripts

Gene Symbol
ACYP1
Entrez Gene ID
97
Gene Name
acylphosphatase 1
Gene Aliases
ACYPE
Chromosome Location
Chr. 14: 75053221 - 75069483 on Build GRCh38
UniGene ID
Hs.18573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107.4 NP_001098.1 3 566
NM_001302616.1 NP_001289545.1 3 548
NM_001302617.1 NP_001289546.1 3 744
NR_126393.1 4 645
NR_126394.1 2 652
NR_126395.1 2 474
GenBank mRNA BC035568.1 - 3 551
BG614847.1 - 4 616
BP316920.1 - 3 529
BX104912.1 - 2 419
X84194.1 - 2 529

View Full Product Details