Gene Transcripts

Gene Symbol
Etnk2
Entrez Gene ID
214253
Gene Name
ethanolamine kinase 2
Gene Aliases
4933417N20Rik, AI197444, EKI 2, Eki2, Tuc1
Chromosome Location
Chr. 1: 133363488 - 133380318 on Build GRCm38
UniGene ID
Mm.52111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175443.5 NP_780652.2 2, 3 715
GenBank mRNA AK031665.1 - 2, 3 501
AK077179.1 - 2, 3 454
AK087818.1 - 1, 2 513
AY212244.1 - 2, 3 306
BC145351.1 - 2, 3 494 9053907
BC152310.1 - 2, 3 653 40089976

View Full Product Details