Gene Transcripts

Gene Symbol
Gpr4
Entrez Gene ID
319197
Gene Name
G protein-coupled receptor 4
Gene Aliases
Chromosome Location
Chr. 7: 19212499 - 19224176 on Build GRCm38
UniGene ID
Mm.135984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175668.4 NP_783599.1 2 2848
GenBank mRNA AK085829.1 - 2 2853

View Full Product Details