Gene Transcripts

Gene Symbol
Rab9b
Entrez Gene ID
319642
Gene Name
RAB9B, member RAS oncogene family
Gene Aliases
9330195C02Rik
Chromosome Location
Chr. X: 136858151 - 136868540 on Build GRCm38
UniGene ID
Mm.44557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176971.2 NP_795945.1 3 2376
GenBank mRNA AK034442.1 - 3 2377
AK049693.1 - 3 2403
AK161867.1 - 3 2598

View Full Product Details