Gene Transcripts

Gene Symbol
Zcchc7
Entrez Gene ID
319885
Gene Name
zinc finger, CCHC domain containing 7
Gene Aliases
4930572I07Rik, D4Wsu132e
Chromosome Location
Chr. 4: 44756559 - 44932215 on Build GRCm38
UniGene ID
Mm.331576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK029808.1 - 6 1398
AK032353.1 - 6 1363
AK039158.1 - 6 1312
AK043238.1 - 6 1304
AK080785.1 - 7 1480
AK157728.1 - 6 1295

View Full Product Details