Gene Transcripts

Gene Symbol
Iqcb1
Entrez Gene ID
320299
Gene Name
IQ calmodulin-binding motif containing 1
Gene Aliases
6820449I09Rik, AV128382, NPHP5
Chromosome Location
Chr. 16: 36828363 - 36872719 on Build GRCm38
UniGene ID
Mm.222934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177128.4 NP_796102.2 14, 15 1744
GenBank mRNA AK078554.1 - 14, 15 1737
AK165220.1 - 14, 15 1728

View Full Product Details