Gene Transcripts

Gene Symbol
CLPTM1
Entrez Gene ID
1209
Gene Name
CLPTM1, transmembrane protein
Gene Aliases
Chromosome Location
Chr. 19: 44954585 - 44993346 on Build GRCh38
UniGene ID
Hs.444441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282175.1 NP_001269104.1 2, 3 201
NM_001282176.1 NP_001269105.1 2, 3 225
NM_001294.3 NP_001285.1 2, 3 352
GenBank mRNA AF037339.1 - 2, 3 195
AK074935.1 - 2, 3 196
AK075262.1 - 2, 3 195
AK223553.1 - 2, 3 196
AK293314.1 - 2, 3 217
AK304480.1 - 2, 3 184
AK310826.1 - 2, 3 225
AK316094.1 - 2, 3 221
BC004865.2 - 2, 3 231 3840991
BC012359.1 - 2, 3 185 4685026
BI916245.1 - 2, 3 198
BT007262.1 - 2, 3 180
DB033341.1 - 2, 3 330
DC313765.1 - 2, 3 201
DC383798.1 - 2, 3 225
HY033360.1 - 2, 3 353
KU177986.1 - 2, 3 180
KU177987.1 - 2, 3 180

View Full Product Details