Gene Transcripts

Gene Symbol
Tnrc6b
Entrez Gene ID
213988
Gene Name
trinucleotide repeat containing 6b
Gene Aliases
2700090M07Rik, A730065C02Rik, AI848765, D230019K20Rik
Chromosome Location
Chr. 15: 80711313 - 80941086 on Build GRCm38
UniGene ID
Mm.131328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144812.2 NP_659061.2 8 1877
NM_177124.4 NP_796098.3 5 1668
GenBank mRNA AK051922.1 - 5 1637
BC068158.1 - 5 1563

View Full Product Details