Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177333.3 | NP_796307.2 | 3 | 280 | |
GenBank mRNA | AK088333.1 | - | 3 | 274 | |
AK089979.1 | - | 3 | 291 | ||
AK131141.1 | - | 3 | 264 | ||
AK141537.1 | - | 3 | 280 | ||
BC031743.1 | - | 3 | 268 | 4187527 |