Gene Transcripts

Gene Symbol
Vwa3a
Entrez Gene ID
233813
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
E030013G06Rik
Chromosome Location
Chr. 7: 120735693 - 120805546 on Build GRCm38
UniGene ID
Mm.389939
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177697.3 NP_808365.2 19 2202
GenBank mRNA AK046643.1 - 1 61
AK086942.1 - 18 1800
AK132732.1 - 19 2203
AK136898.1 - 18 1801

View Full Product Details