Gene Transcripts
- Gene Symbol
- Ubr3
- Entrez Gene ID
- 68795
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 3
- Gene Aliases
- 1110059H15Rik, 4833421P10Rik, A130030D10Rik, AA409735, AA414972, AA422631, AI646861, AU016126, Zfp650, Znf650
- Chromosome Location
- Chr. 2: 69896969 - 70024010 on Build GRCm38
- UniGene ID
- Mm.314576
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037622.1 |
- |
2, 3 |
671 |
|
View Full Product Details