Gene Transcripts

Gene Symbol
Slc7a15
Entrez Gene ID
328059
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 15
Gene Aliases
Arpat
Chromosome Location
Chr. 12: 8528481 - 8599066 on Build GRCm38
UniGene ID
Mm.46326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038660.2 NP_001033749.1 6 1639
NM_177802.3 NP_808470.1 7 1196
GenBank mRNA AK033481.1 - 7 1196

View Full Product Details